Novus Biologicals
Manufacturer Code:NB1006990.1ML
Catalog # NB017794
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AMT28 |
Immunogen | Prokaryotic recombinant protein corresponding to the 180 amino acid N-terminal region of the human CDX2 molecule. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: caudal type homeo box transcription factor 2; caudal type homeo box transcription factor 2 caudal type homeobox 2 Caudal-type homeobox protein 2 CDX-3 CDX3caudal type homeobox transcription factor 2 homeobox protein CDX-2; caudal type homeobox transcription factor 2; Caudal-type homeobox protein 2; CDX-3; Homeobox protein CDX-2; homeobox protein miniCDX2
Gene Aliases: CDX-3; CDX2; CDX2/AS; CDX3
UniProt ID: (Human) Q99626
Entrez Gene ID: (Human) 1045
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.