Proteintech
Manufacturer Code:143821AP150UL
Catalog # 50-172-9413
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CDT1 Fusion Protein Ag5810 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDT1; CDT1 DNA replication factor Cdt1 Double parked homolog DUP RIS2; DNA replication factor Cdt1; Double parked homolog; Double parked, Drosophila, homolog of; DUP
Gene Aliases: CDT1; DUP; RIS2
UniProt ID: (Human) Q9H211
Entrez Gene ID: (Human) 81620
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.