Invitrogen
Manufacturer Code:PA590918
Catalog # PIPA590918
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CDS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDP-DAG synthase 2; CDP-DG synthase 2; CDP-diacylglycerol synthase 2; CDP-diglyceride pyrophosphorylase 2; CDP-diglyceride synthase 2; CDS 2; CTP:phosphatidate cytidylyltransferase 2; Phosphatidate cytidylyltransferase 2
Gene Aliases: 5730450N06Rik; 5730460C18Rik; AI854580; Cds2; D2Wsu127e
UniProt ID: (Mouse) Q99L43
Entrez Gene ID: (Mouse) 110911
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.