Proteintech
Manufacturer Code:176341AP150UL
Catalog # 50-173-1695
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CDON Fusion Protein Ag11904 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAM-related/down-regulated by oncogenes; CDO CDON Cdon homolog (mouse) ORCAM; Cdon homolog; Cell adhesion molecule-related/down-regulated by oncogenes; immunoglobulin superfamily member; immunoglobulin superfamily member CDO; oncogene-regulated cell adhesion molecule; surface glycoprotein, Ig superfamily member
Gene Aliases: Cd0; CDO; CDON; CDON1; HPE11; ORCAM
UniProt ID: (Human) Q4KMG0, (Mouse) Q32MD9, (Rat) O35158
Entrez Gene ID: (Human) 50937, (Mouse) 57810, (Rat) 50938
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.