Invitrogen
Manufacturer Code:PA538005
Catalog # PIPA538005
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | 19 amino acid peptide near the amino terminus of human CDO1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDO; CDO-I; cysteine dioxygenase 1, cytosolic; Cysteine dioxygenase type 1; Cysteine dioxygenase type I; Cysteine dioxygenase type I CDO CDO-I CDOI; cysteine dioxygenase, type I; cytosolic cysteine dioxygenase 1
Gene Aliases: 1300002L19Rik; Cdo; CDO-I; CDO1; D18Ucla3
UniProt ID: (Human) Q16878, (Rat) P21816, (Mouse) P60334
Entrez Gene ID: (Human) 1036, (Rat) 81718, (Mouse) 12583
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.