R&D Systems
Manufacturer Code:AF2429
Catalog # AF2429
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse CDO Val22-Leu963 Accession # AAC43031 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CDON Cdon homolog (mouse) CDOORCAM cell adhesion molecule-related/down-regulated by oncogenes MGC111524 ORCAM surface glycoprotein Ig superfamily member; Cdon homolog; Cell adhesion molecule-related/down-regulated by oncogenes; surface glycoprotein, Ig superfamily member
Gene Aliases: CDO; CDON; CDON1; HPE11; ORCAM
UniProt ID: (Human) Q4KMG0
Entrez Gene ID: (Human) 50937
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.