OriGene
Manufacturer Code:TA802082
Catalog # 50-167-9101
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5H8 |
Immunogen | Full length human recombit protein of human CDKN2A produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Alternative reading frame; ARF; ARF CDK4I CDKN2 CMM2 INK4 INK4A MLM MTS-1 MTS1 P14 P14ARF P16 P16-INK4A P16INK4 P16INK4A P19 P19ARF TP16; CDK4 inhibitor p16-INK4; CDK4I; cell cycle negative regulator beta; Cyclin-dependent kinase 4 inhibitor A; Cyclin-dependent kinase inhibitor 2A; cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4); MTS-1; Multiple tumor suppressor 1; p14ARF; p16-INK4; p16-INK4a; Tumor suppressor ARF
Gene Aliases: ARF; CDK4I; CDKN2; CDKN2A; CMM2; INK4; INK4A; MLM; MTS-1; MTS1; P14; P14ARF; P16; P16-INK4A; P16INK4; P16INK4A; P19; P19ARF; TP16
UniProt ID: (Human) P42771
Entrez Gene ID: (Human) 1029
Molecular Function: enzyme modulator kinase inhibitor kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.