Abnova
Manufacturer Code:MAB20863
Catalog # 89-337-360
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CIP1/823 &plus DCS-60.2 |
Immunogen | Recombinant protein corresponding to full length human CDKN1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 2 to 8°C |
Mouse Monoclonal Antibody.
Protein Aliases: CAP20 CDKN1 CIP1 MDA-6 P21 SDI1 WAF1 p21CIP1; CDK-interacting protein 1; CDK-interaction protein 1; Cyclin-dependent kinase inhibitor 1; cyclin-dependent kinase inhibitor 1A (p21, Cip1); DNA synthesis inhibitor; MDA-6; melanoma differentiation associated protein 6; Melanoma differentiation-associated protein 6; p21; wild-type p53-activated fragment 1
Gene Aliases: CAP20; CDKN1; CDKN1A; CIP1; MDA-6; MDA6; P21; p21CIP1; PIC1; SDI1; WAF1
UniProt ID: (Human) P38936
Entrez Gene ID: (Human) 1026
Molecular Function: enzyme modulator kinase inhibitor kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.