Invitrogen
Manufacturer Code:PA585238
Catalog # PIPA585238
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human CDK9. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-2K; C-2k antibody CDC2L4 antibody CTK1 antibody PITALRE antibody TAK antibody CDK9 antibody serine/threonine-protein kinase PITALRE antibody cyclin-dependent kinase 9 antibody serine/threonine protein kinase PITALRE antibody CDC2-related kinase antibody cell division cycle 2-like protein kinase 4 antibody cell division protein kinase 9 antibody tat-associated kinase complex catalytic subunit antibody; CDC2-related kinase; Cell division cycle 2-like protein kinase 4; Cell division protein kinase 9; Cyclin-dependent kinase 9; cyclin-dependent kinase 9 (CDC2-related kinase); serine/threonine protein kinase PITALRE; Serine/threonine-protein kinase PITALRE; Tat-associated kinase complex catalytic subunit
Gene Aliases: C-2k; CDC2L4; CDK9; CTK1; PITALRE; TAK
UniProt ID: (Human) P50750, (Mouse) Q3UQI6, (Rat) Q641Z4
Entrez Gene ID: (Human) 1025, (Mouse) 107951, (Rat) 362110
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.