Proteintech
Manufacturer Code:117051AP150UL
Catalog # 50-172-7389
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CDK9 Fusion Protein Ag2318 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C 2k CDC2L4 CDK9 Cell division protein kinase 9 CTK1 cyclin dependent kinase 9 PITALRE TAK; C-2K; CDC2-related kinase; Cell division cycle 2-like protein kinase 4; Cell division protein kinase 9; Cyclin-dependent kinase 9; cyclin-dependent kinase 9 (CDC2-related kinase); serine/threonine protein kinase PITALRE; Serine/threonine-protein kinase PITALRE; Tat-associated kinase complex catalytic subunit
Gene Aliases: C-2k; CDC2L4; CDK9; CTK1; PITALRE; TAK
UniProt ID: (Human) P50750, (Mouse) Q3UQI6, (Rat) Q641Z4
Entrez Gene ID: (Human) 1025, (Mouse) 107951, (Rat) 362110
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.