Abnova
Manufacturer Code:PAB25914
Catalog # 89-127-544
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding Y13 of human CDK6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CDK6.
For Research Use Only
Protein Aliases: Cell division protein kinase 6; Cyclin-dependent kinase 6; MGC59692; PLSTIRE; Serine/threonine-protein kinase PLSTIRE; STQTL11
Gene Aliases: CDK6; CDKN6; MCPH12; PLSTIRE
UniProt ID: (Human) Q00534
Entrez Gene ID: (Human) 1021
Molecular Function: kinase non-receptor serine/threonine protein kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.