Invitrogen
Manufacturer Code:PA538872
Catalog # PIPA538872
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human CDK5RAP3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDK5 activator-binding protein C53; CDK5 activator-binding protein C53; CDK5 regulatory subunit associated protein IC53-2; CDK5 regulatory subunit-associated protein 3; IC53-2; ischemic heart CDK5 activator-binding protein C53; LXXLL/leucine-zipper-containing ARF-binding protein; LXXLL/leucine-zipper-containing ARFbinding protein; Protein HSF-27
Gene Aliases: 1810007E24Rik; BC002318; C53; C81486; CDK5RAP3; HSF-27; IC53; LZAP; MST016; MSTP016; OK/SW-cl.114; PP1553
UniProt ID: (Human) Q96JB5, (Mouse) Q99LM2
Entrez Gene ID: (Human) 80279, (Rat) 80278, (Mouse) 80280
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.