Invitrogen
Manufacturer Code:PA5115000
Catalog # PIPA5115000
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CDK5RAP3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1810007E24Rik BC002318 C53 C81486 CDK5 activator-binding protein C53 CDK5 activator-binding protein C53 CDK5 regulatory subunit associated protein 3 CDK5 regulatory subunit associated protein IC53-2 CDK5 regulatory subunit-associated protein 3 CDK5RAP3 HSF-27 IC53 IC53-2 ischemic heart CDK5 activator-binding protein C53 LXXLL/leucine-zipper-containing ARFbinding protein LXXLL/leucine-zipper-containing ARF-binding protein LZAP MST016 MSTP016 OK/SW-cl.114 PP1553 Protein HSF-27; CDK5 activator-binding protein C53; CDK5 activator-binding protein C53; CDK5 regulatory subunit associated protein IC53-2; CDK5 regulatory subunit-associated protein 3; ischemic heart CDK5 activator-binding protein C53; LXXLL/leucine-zipper-containing ARF-binding protein; LXXLL/leucine-zipper-containing ARFbinding protein; Protein HSF-27
Gene Aliases: 1810007E24Rik; BC002318; C53; C81486; CDK5RAP3; HSF-27; IC53; LZAP; MST016; MSTP016; OK/SW-cl.114; PP1553
UniProt ID: (Human) Q96JB5, (Mouse) Q99LM2
Entrez Gene ID: (Human) 80279, (Rat) 80278, (Mouse) 80280
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.