Novus Biologicals
Manufacturer Code:NBP186778
Catalog # NBP186778
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:GDFGVEAVSEGTDSGISAEAAGIDWGIFPESDSKDPGGDGIDWGDDAVALQITVLEAGTQAPEGVARGPDALTLLEYTETRNQFLDELMELEIFLAQRAVELSEEADVLSVSQFQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C53 CDK5 activator-binding protein C53 CDK5 regulatory subunit associated protein 3 FLJ13660 HSF-27 IC53CDK5 regulatory subunit-associated protein 3 ischemic heart CDK5 activator-binding protein C53 LXXLL/leucine-zipper-containing ARFbinding protein LZAP MST016 OK/SW-cl.114 Protein HSF-27; CDK5 activator-binding protein C53; CDK5 regulatory subunit associated protein IC53-2; CDK5 regulatory subunit-associated protein 3; ischemic heart CDK5 activator-binding protein C53; LXXLL/leucine-zipper-containing ARF-binding protein; LXXLL/leucine-zipper-containing ARFbinding protein; Protein HSF-27
Gene Aliases: C53; CDK5RAP3; HSF-27; IC53; LZAP; MST016; MSTP016; OK/SW-cl.114; PP1553
UniProt ID: (Human) Q96JB5
Entrez Gene ID: (Human) 80279
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.