Invitrogen
Manufacturer Code:711426
Catalog # PI711426
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Oligoclonal |
Type | Antibody |
Clone | 13HCLC |
Immunogen | Protein corresponding to Human CDK5RAP2 (aa 1-300) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Polyclonal Antibody
For Research Use Only
Protein Aliases: CDK5 activator-binding protein C48; CDK5 regulatory subunit-associated protein 2; centrosomal protein 215 kDa; Centrosome-associated protein 215; centrosomin
Gene Aliases: C48; CDK5RAP2; CEP215; KIAA1633; MCPH3
UniProt ID: (Human) Q96SN8
Entrez Gene ID: (Human) 55755
Molecular Function:
G-protein modulator
actin binding motor protein
actin family cytoskeletal protein
cell junction protein
cytoskeletal protein
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.