Abnova
Manufacturer Code:PAB24886
Catalog # 89-126-557
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to CDK5R2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of CDK5R2.
For Research Use Only
Protein Aliases: CDK5 activator 2; Cyclin-dependent kinase 5 activator 2; cyclin-dependent kinase 5 activator isoform p39i; Cyclin-dependent kinase 5 regulatory subunit 2; cyclin-dependent kinase 5, regulatory subunit 2 (p39); NCK5AI; neuronal CDK5 activator isoform; P39; p39I; p39nck5ai
Gene Aliases: CDK5R2; NCK5AI; P39; p39nck5ai
UniProt ID: (Human) Q13319
Entrez Gene ID: (Human) 8941
Molecular Function:
enzyme modulator
kinase activator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.