Novus Biologicals
Manufacturer Code:NBP200776
Catalog # NBP200776
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2D9 |
Immunogen | Full length human recombinant protein of human CDK2 (NP_001789) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cdc2-related protein kinase; cdc2-related protein kinase cell devision kinase 2 Cell division protein kinase 2 cyclin-dependent kinase 2 EC 2.7.11 EC 2.7.11.22 p33 protein kinase p33(CDK2); Cell division protein kinase 2; Cyclin-dependent kinase 2; p33 protein kinase
Gene Aliases: CDK2; CDKN2; p33(CDK2)
UniProt ID: (Human) P24941
Entrez Gene ID: (Human) 1017
Molecular Function: kinase non-receptor serine/threonine protein kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.