R&D Systems
Manufacturer Code:AF4654SP
Catalog # AF4654SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human CDK2 Met1-Leu298 Accession # P24941 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: cdc2-related protein kinase; cdc2-related protein kinase cell devision kinase 2 Cell division protein kinase 2 cyclin-dependent kinase 2 EC 2.7.11 EC 2.7.11.22 p33 protein kinase p33(CDK2); Cell division protein kinase 2; Cyclin-dependent kinase 2; p33 protein kinase
Gene Aliases: CDK2; CDKN2; p33(CDK2)
UniProt ID: (Human) P24941
Entrez Gene ID: (Human) 1017
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.