Invitrogen
Manufacturer Code:PA526054
Catalog # PIPA526054
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 99-125 amino acids from the central region of human CDIPT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDP-diacylglycerol--inositol 3-phosphatidyltransferase; Phosphatidylinositol synthase; PI synthase; PIS PIS1 PI synthase PtdIns synthase phosphatidylinositol synthase; PtdIns synthase
Gene Aliases: 9530042F15Rik; CDIPT; D7Bwg0575e; PIS; PIS1
UniProt ID: (Human) O14735, (Mouse) Q8VDP6
Entrez Gene ID: (Human) 10423, (Mouse) 52858
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.