Novus Biologicals
Manufacturer Code:NBP2826430.1ML
Catalog # NB030572
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C terminal region of human CDIPT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDP-diacylglycerol--inositol 3-phosphatidyltransferase; CDP-diacylglycerol--inositol 3-phosphatidyltransferase EC 2.7.8.11 Phosphatidylinositol synthase PI synthase PIS1MGC1328 PISCDP-diacylglycerol--inositol 3-phosphatidyltransferase (phosphatidylinositolsynthase) PtdIns synthase; Phosphatidylinositol synthase; PI synthase; PtdIns synthase
Gene Aliases: CDIPT; PIS; PIS1
UniProt ID: (Human) O14735
Entrez Gene ID: (Human) 10423
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.