Proteintech
Manufacturer Code:134961AP150UL
Catalog # 50-172-8783
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CDH23 Fusion Protein Ag4322 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: age related hearing loss 1; bobby; bustling; CAD23 Cadherin 23 cadherin like 23 CDH23 DFNB12 KIAA1774 KIAA1812 Otocadherin USH1D; cadherin 23 (otocadherin); cadherin related 23; Cadherin-23; cadherin-like 23; cadherin-related family member 23; modifier of deaf waddler; Otocadherin; waltzer; Waltzing
Gene Aliases: 4930542A03Rik; ahl; ahl1; bob; bus; CDH23; CDHR23; KIAA1774; KIAA1812; mdfw; nmf112; nmf181; nmf252; sals; UNQ1894/PRO4340; USH1D; v; W
UniProt ID: (Human) Q9H251, (Mouse) Q99PF4, (Rat) P58365
Entrez Gene ID: (Human) 64072, (Mouse) 22295, (Rat) 114102
Molecular Function: cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.