OriGene
Manufacturer Code:CF503933
Catalog # 50-166-2494
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2G7 |
Immunogen | Full length human recombit protein of human CDH2 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cadherin 2, type 1, N-cadherin (neuronal); Cadherin-2; calcium-dependent adhesion protein, neuronal; CD325; CD325 CDHN CDw325 NCAD; CDw325; N-cadherin; N-cadherin 1; Neural cadherin
Gene Aliases: CD325; CDH2; CDHN; CDw325; N-cadherin; NCAD
UniProt ID: (Human) P19022, (Mouse) P15116
Entrez Gene ID: (Dog) 480169, (Human) 1000, (Mouse) 12558
Molecular Function: cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.