Abnova
Manufacturer Code:PAB7876
Catalog # 89-133-457
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues surrounding Y820 of human CDH2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of CDH2.
For Research Use Only
Protein Aliases: cadherin 2, type 1, N-cadherin (neuronal); Cadherin-2; calcium-dependent adhesion protein, neuronal; CD325; CDHN; CDw325; N-cadherin; N-cadherin 1; NCAD; Neural cadherin
Gene Aliases: CD325; CDH2; CDHN; CDw325; NCAD
UniProt ID: (Human) P19022
Entrez Gene ID: (Human) 1000
Molecular Function: cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.