Invitrogen
Manufacturer Code:MA518086
Catalog # PIMA518086
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 16A6 |
Immunogen | Extracellular domain of human cadherin-11 fused to maltose binding protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cadherin 11, type 2, OB-cadherin (osteoblast); Cadherin-11; OB Cadherin Cadherin 11 Osteoblast Cadherin OSF-4; OB-cadherin; OSF-4; Osteoblast cadherin
Gene Aliases: CAD11; CDH11; CDHOB; OB; OSF-4
UniProt ID: (Human) P55287
Entrez Gene ID: (Human) 1009
Molecular Function:
cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.