Invitrogen
Manufacturer Code:PA596937
Catalog # PIPA596937
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 637-796 of human CDH11. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cadherin 11, type 2, OB-cadherin (osteoblast); Cadherin-11; OB-cadherin; OSF-4; Osteoblast cadherin; osteoblast-cadherin
Gene Aliases: Cad-11; CAD11; CDH11; CDHOB; OB; OSF-4
UniProt ID: (Human) P55287, (Mouse) P55288
Entrez Gene ID: (Human) 1009, (Rat) 84407, (Mouse) 12552
Molecular Function:
cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.