Abnova
Manufacturer Code:PAB19043
Catalog # 89-120-598
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 497-509 at internal region of human CDH11. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of CDH11.
For Research Use Only
Protein Aliases: CAD11; cadherin 11, type 2, OB-cadherin (osteoblast); Cadherin-11; CDHOB; OB; OB-cadherin; OSF-4; Osteoblast cadherin
Gene Aliases: CAD11; CDH11; CDHOB; OB; OSF-4
UniProt ID: (Human) P55287
Entrez Gene ID: (Human) 1009
Molecular Function:
cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.