OriGene
Manufacturer Code:UM870076
Catalog # 50-168-5526
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | UMAB184 |
Immunogen | The extracellular domain of human CDH1 produced in human cells. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Arc-1 BCDS1 CD324 CDHE ECAD LCAM UVO; cadherin 1, E-cadherin (epithelial); cadherin 1, type 1; cadherin 1, type 1, E-cadherin (epithelial); Cadherin-1; calcium-dependent adhesion protein, epithelial; CAM 120/80; CD324; cell-CAM 120/80; E-Cad/CTF1; E-Cad/CTF2; E-Cad/CTF3; E-Cadherin; E-cadherin 1; Epithelial cadherin; Uvomorulin
Gene Aliases: Arc-1; CD324; CDH1; CDHE; ECAD; LCAM; UVO
UniProt ID: (Human) P12830
Entrez Gene ID: (Human) 999
Molecular Function:
cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.