Invitrogen
Manufacturer Code:PA541747
Catalog # PIPA541747
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human CDC5L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDC5 cell division cycle 5-like; CDC5 cell division cycle 5-like (S. pombe) CEF1 KIAA0432 PCDC5RP dJ319D22.1 hCDC5 CDC5 CDC5-LIKE; Cdc5-like protein; Cdc5-related protein; Cell division cycle 5-like protein; cell division cycle 5-related protein; dJ319D22.1 (CDC5-like protein); Pombe Cdc5-related protein
Gene Aliases: 1200002I02Rik; AA408004; CDC5; CDC5-LIKE; CDC5L; CEF1; dJ319D22.1; KIAA0432; PCDC5RP; zgc:55853
UniProt ID: (Human) Q99459, (Bovine) Q2KJC1, (Rat) O08837, (Mouse) Q6A068
Entrez Gene ID: (Horse) 100055574, (Dog) 474921, (Human) 988, (Goat) 100861007, (Rabbit) 100340843, (Bovine) 767817, (Rat) 85434, (Guinea pig) 100729105, (Mouse) 71702, (Zebrafish) 394059
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.