Novus Biologicals
Manufacturer Code:NB10087014
Catalog # NB10087014
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 752 and 802 of human cell division cycle 5 (CDC5) -like using the numbering given in entry NP_001244.1 (GeneID988). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDC5 CDC5 (cell division cycle 5 S. pombe homolog)-like CDC5 cell division cycle 5-like (S. pombe) Cdc5-like protein Cdc5-related protein CEF1 cell division cycle 5-like protein dJ319D22.1 dJ319D22.1 (CDC5-like protein) hCDC5 KIAA0432 PCDC5RPCDC5-LIKE Pombe cdc5-related protein; CDC5 cell division cycle 5-like; Cdc5-like protein; Cdc5-related protein; Cell division cycle 5-like protein; dJ319D22.1 (CDC5-like protein); Pombe cdc5-related protein
Gene Aliases: CDC5; CDC5-LIKE; CDC5L; CEF1; dJ319D22.1; KIAA0432; PCDC5RP
UniProt ID: (Human) Q99459
Entrez Gene ID: (Human) 988
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.