Abnova
Manufacturer Code:H00000988K
Catalog # 89-205-537
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human CDC5L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human CDC5L peptide using ARM Technology.
For Research Use Only
Protein Aliases: CDC5 cell division cycle 5-like; Cdc5-like protein; Cdc5-related protein; CEF1 KIAA0432 PCDC5RP dJ319D22.1 hCDC5; Cell division cycle 5-like protein; dJ319D22.1 (CDC5-like protein); Pombe cdc5-related protein
Gene Aliases: CDC5; CDC5-LIKE; CDC5L; CEF1; dJ319D22.1; KIAA0432; PCDC5RP
UniProt ID: (Human) Q99459
Entrez Gene ID: (Human) 988
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.