Novus Biologicals
Manufacturer Code:NBP214907
Catalog # NBP214907
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human CDC42. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDC42Hs cell division control protein 42 homolog cell division cycle 42 (GTP binding protein 25kDa) cell division cycle 42 (GTP-binding protein 25kD) dJ224A6.1.1 (cell division cycle 42 (GTP-binding protein 25kD)) dJ224A6.1.2 (cell division cycle 42 (GTP-binding protein 25kD)) G25K G25K GTP-binding protein growth-regulating protein GTP-binding protein 25kD small GTP binding protein CDC42; Cell division control protein 42 homolog; dJ224A6.1.1 (cell division cycle 42 (GTP-binding protein, 25kD)); dJ224A6.1.2 (cell division cycle 42 (GTP-binding protein, 25kD)); G25K GTP-binding protein; growth-regulating protein; GTP binding protein, 25kDa; small GTP binding protein CDC42
Gene Aliases: CDC42; CDC42Hs; G25K; TKS
UniProt ID: (Human) P60953
Entrez Gene ID: (Human) 998
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.