Novus Biologicals
Manufacturer Code:NBP298725H
Catalog # NB055006
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant CDC42 (P60953-2 Met1-Cys188). |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDC42Hs cell division control protein 42 homolog cell division cycle 42 (GTP binding protein 25kDa) cell division cycle 42 (GTP-binding protein 25kD) dJ224A6.1.1 (cell division cycle 42 (GTP-binding protein 25kD)) dJ224A6.1.2 (cell division cycle 42 (GTP-binding protein 25kD)) G25K G25K GTP-binding protein growth-regulating protein GTP-binding protein 25kD small GTP binding protein CDC42; Cell division control protein 42 homolog; dJ224A6.1.1 (cell division cycle 42 (GTP-binding protein, 25kD)); dJ224A6.1.2 (cell division cycle 42 (GTP-binding protein, 25kD)); G25K GTP-binding protein; growth-regulating protein; GTP binding protein, 25kDa; small GTP binding protein CDC42
Gene Aliases: CDC42; CDC42Hs; G25K; TKS
UniProt ID: (Human) P60953
Entrez Gene ID: (Human) 998
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.