Proteintech
Manufacturer Code:162931AP150UL
Catalog # 50-173-0858
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CDC37L1 Fusion Protein Ag9459 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDC37 cell division cycle 37 homolog-like 1; CDC37B CDC37L1 HARC; cell division cycle 37 homolog-like 1; cell division cycle 37-like 1; Hsp90 co-chaperone Cdc37-like 1; Hsp90-associating relative of Cdc37
Gene Aliases: CDC37B; CDC37L1; HARC
UniProt ID: (Human) Q7L3B6
Entrez Gene ID: (Human) 55664
Molecular Function: chaperone enzyme modulator kinase activator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.