Novus Biologicals
Manufacturer Code:NBP215827
Catalog # NBP215827
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human CDC25C. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDC 25 CDC25 cell division cycle 25 homolog C (S. cerevisiae) cell division cycle 25 homolog C (S. pombe) cell division cycle 25C Dual specificity phosphatase Cdc25C EC 3.1.3.48 mitosis inducer CDC25 M-phase inducer phosphatase 3 phosphotyrosine phosphatase; Dual specificity phosphatase Cdc25C; M-phase inducer phosphatase 3; mitosis inducer CDC25; phosphotyrosine phosphatase; protein phosphatase 1, regulatory subunit 60
Gene Aliases: CDC25; CDC25C; PPP1R60
UniProt ID: (Human) P30307
Entrez Gene ID: (Human) 995
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.