Proteintech
Manufacturer Code:106441AP150UL
Catalog # 50-172-6474
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CDC25B Fusion Protein Ag0994 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDC25B CDC25HU2 M phase inducer phosphatase 2; Dual specificity phosphatase Cdc25B; M-phase inducer phosphatase 2
Gene Aliases: AI604853; CDC25B; CDC25HU2; Cdc25m2
UniProt ID: (Human) P30305, (Mouse) P30306
Entrez Gene ID: (Human) 994, (Mouse) 12531
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.