Proteintech
Manufacturer Code:550311AP150UL
Catalog # 50-173-5945
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDC25 isoform A1-CAG; CDC25A CDC25A2 M phase inducer phosphatase 1; CDC25A2-CAG isoform; Dual specificity phosphatase Cdc25A; M-phase inducer phosphatase 1
Gene Aliases: CDC25A; CDC25A2; Cdc25m3; D9Ertd393e
UniProt ID: (Human) P30304, (Mouse) P48964
Entrez Gene ID: (Human) 993, (Mouse) 12530
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.