Invitrogen
Manufacturer Code:MA146040
Catalog # PIMA146040
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | BA8 |
Immunogen | Synthetic C terminal peptide (Xenopus laevis). BLAST &primeFEVDPVTKKEKEKARSSKSIIHQSIR&prime |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cell division cycle 20 homolog
Gene Aliases: cdc20; fizzy1; X-FZY; XELAEV_18025163mg
Entrez Gene ID: (Xenopus) 379130
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.