Proteintech
Manufacturer Code:136601AP150UL
Catalog # 50-172-8916
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CDC14A Fusion Protein Ag4556 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDC10 (cell division cycle 10, S. cerevisiae, homolog); cdc14 CDC14A hCDC14; CDC14 cell division cycle 14 homolog A; Dual specificity protein phosphatase CDC14A
Gene Aliases: A830059A17Rik; Cdc14; CDC14A; CDC14a1; CDC14A2; DFNB105; hCDC14
UniProt ID: (Human) Q9UNH5, (Mouse) Q6GQT0
Entrez Gene ID: (Human) 8556, (Rat) 310806, (Mouse) 229776
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.