Invitrogen
Manufacturer Code:PA5102545
Catalog # PIPA5102545
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CDA corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2210401N16Rik Cda Cdd Cytidine aminohydrolase cytidine deaminase cytosine nucleoside deaminase EC 3.5.4.5 small cytidine deaminase; Cytidine aminohydrolase; Cytidine deaminase; cytosine nucleoside deaminase; small cytidine deaminase
Gene Aliases: 2210401N16Rik; CDA; CDD
UniProt ID: (Human) P32320, (Mouse) P56389
Entrez Gene ID: (Human) 978, (Mouse) 72269, (Rat) 362638
Molecular Function:
deaminase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.