R&D Systems
Manufacturer Code:MAB5920
Catalog # MAB5920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 590559 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human CD98 Asp105-Gln529 Accession # P08195 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 4F2 4F2hc 4T2HC antigen identified by monoclonal antibodies 4F2 TRA1.10 TROP4 and T434F2HC CD98 CD98 antigen CD98 heavy chain CD98HC CH98hc FRP-1 heavy chain Lymphocyte activation antigen 4F2 large subunit MDU1antigen defined by monoclonal 4F2 heavy chain monoclonal 44D7 NACAE4F2 cell-surface antigen heavy chain SLC3A2 solute carrier family 3 (activators of dibasic and neutral amino acidtransport) member 24F2 heavy chain antigen; 4F2 cell-surface antigen heavy chain; 4F2 heavy chain antigen; 4F2hc; antigen defined by monoclonal antibody 4F2, heavy chain; antigen identified by monoclonal antibodies 4F2, TRA1.10, TROP4, and T43; CD98; CD98 heavy chain; Lymphocyte activation antigen 4F2 large subunit; monoclonal antibody 44D7; solute carrier family 3 (activators of dibasic and neutral amino acid transport), member 2; solute carrier family 3 (amino acid transporter heavy chain), member 2; Solute carrier family 3 member 2
Gene Aliases: 4F2; 4F2HC; 4T2HC; CD98; CD98HC; MDU1; NACAE; SLC3A2
UniProt ID: (Human) P08195
Entrez Gene ID: (Human) 6520
Molecular Function: amylase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.