R&D Systems
Manufacturer Code:AF2067SP
Catalog # AF2067SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human CD90/Thy1 Gln20-Ser141 Accession # P04216 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: CD90; CD90 CD90 antigen CDw90 FLJ33325 Thy1 Thy-1 antigen Thy-1 cell surface antigen thy-1 membrane glycoprotein Thy-1 T-cell antigen; CDw90; Thy-1 antigen; Thy-1 membrane glycoprotein; Thy-1 T-cell antigen
Gene Aliases: CD90; CDw90; THY1
UniProt ID: (Human) P04216
Entrez Gene ID: (Human) 7070
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.