Proteintech
Manufacturer Code:667661IG150UL
Catalog # 50-173-7172
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2D7D11 |
Immunogen | CD90 Fusion Protein Ag25603 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD90; CD90 CDw90 FLJ33325 Thy 1 antigen Thy 1 cell surface antigen Thy 1 membrane glycoprotein THY1; CDw90; membrane glycoprotein; Thy-1 antigen; thy-1 glycoprotein; Thy-1 membrane glycoprotein; Thy-1 protein; Thy-1 T-cell antigen; thymus cell antigen 1, theta; Thymus cell surface antigen
Gene Aliases: CD7; CD90; CDw90; Thy-1; THY1
UniProt ID: (Human) P04216, (Rat) P01830
Entrez Gene ID: (Human) 7070, (Pig) 100271931, (Rat) 24832
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.