Invitrogen
Manufacturer Code:15086281
Catalog # 50-140-78
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | GL1 |
Conjugate | PE-Cyanine5 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Activation B7-2 antigen; B7-2 B7.2 B70 CD28LG2 LAB72 MGC34413 B-lymphocyte activation antigen B7-2 B-lymphocyte antigen B7-2 B7-2 antigen BU63 CD28 antigen ligand 2; CD86; early T cell costimulatory molecule-1; Early T-cell costimulatory molecule 1; ETC-1; T-lymphocyte activation antigen CD86
Gene Aliases: B7; B7-2; B7.2; B70; Cd28l2; Cd86; CLS1; ETC-1; Ly-58; Ly58; MB7; MB7-2; TS/A-2
UniProt ID: (Mouse) P42082
Entrez Gene ID: (Mouse) 12524
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.