TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | 5 µL/1x10^6 cells |
PUBLISHED APPLICATIONS | |
---|---|
Flow Cytometry (Flow) | See 1 publications below |
Published species | Human |
Host / Isotype | Mouse / IgG1 |
Class | Monoclonal |
Type | Antibody |
Clone | BU63 |
Immunogen | B-lymphoblastoid cell line ARH 77 |
Conjugate |
Alexa Fluor™ 488
|
Form | Liquid |
Concentration | 0.12 mg/mL |
Storage conditions | 4° C, store in dark |
RRID | AB_2633292 |
CD86 is one of two ligands (the other CD80) for CTLA4 and CD28. CD86 acts as costimulatory molecule in eliciting T-cell help during antigen presentation. Antigen presentation in the absence of sufficient co-stimulation involving CD86/CD80 can induce tolerance. CD80 appears to play a role distinct from CD80 in T helper cell differentiation. CD86 is a type I membrane protein that is a member of the immunoglobulin superfamily. The CD86 protein is expressed by antigen-presenting cells, and it is the ligand for two proteins at the cell surface of T cells, CD28 antigen and cytotoxic T-lymphocyte-associated protein 4. Binding of CD86 with CD28 antigen is a costimulatory signal for activation of the T-cell. Binding of CD86 with cytotoxic T-lymphocyte-associated protein 4 negatively regulates T-cell activation and diminishes the immune response. Alternative splicing results in two transcript variants encoding different isoforms of CD86. Additional transcript variants have been described for CD86, but their full-length sequences have not been determined. Diseases associated with CD86 dysfunction include gallbladder squamous cell carcinoma and myocarditis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.