Invitrogen
Manufacturer Code:MA135969
Catalog # 01672454
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | BU63 |
Immunogen | Human CD86. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4°C or -20°C if preferred store in dark |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Activation B7-2 antigen; B-lymphocyte activation antigen B7-2; B7-2 B7.2 B70 CD28LG2 LAB72 MGC34413 B-lymphocyte activation antigen B7-2 B-lymphocyte antigen B7-2 B7-2 antigen BU63 CD28 antigen ligand 2; B70; BU63; CD86; CD86 antigen (CD28 antigen ligand 2, B7-2 antigen); CTLA-4 counter-receptor B7.2; FUN-1; T-lymphocyte activation antigen CD86
Gene Aliases: B7-2; B7.2; B70; CD28LG2; CD86; LAB72
UniProt ID: (Human) P42081
Entrez Gene ID: (Human) 942
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.