Abnova
Manufacturer Code:MAB10242
Catalog # 89-108-447
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CD84.1.21 |
Immunogen | CD84-transfected 300.19 cell line. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Mouse monoclonal antibody raised against of CD84.
For Research Use Only
Protein Aliases: CD84; CD84 antigen (leukocyte antigen); Cell surface antigen MAX.3; DKFZp781E2378; hCD84; Hly9-beta; leucocyte differentiation antigen CD84; leukocyte antigen CD84; Leukocyte differentiation antigen CD84; LY9B; mCD84; Signaling lymphocytic activation molecule 5; SLAM family member 5; SLAMF5
Gene Aliases: CD84; hCD84; LY9B; mCD84; SLAMF5
UniProt ID: (Human) Q9UIB8
Entrez Gene ID: (Human) 8832
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
kinase
protein kinase
receptor
signaling molecule
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.