Invitrogen
Manufacturer Code:MA110292
Catalog # PIMA110292
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | M38 |
Immunogen | MOLT-4 (human T-ALL cell line) |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 26 kDa cell surface protein TAPA-1; 26 kDa cell surface protein TAPA-1 CD81 antigen CD81 antigen target of antiproliferative antibody 1 tetraspanin-28 tspan-28; CD81; CD81 antigen; CD81 antigen (target of antiproliferative antibody 1); Target of the antiproliferative antibody 1; Tetraspanin-28; Tspan-28
Gene Aliases: CD81; CVID6; S5.7; TAPA1; TSPAN28
UniProt ID: (Human) P60033
Entrez Gene ID: (Human) 975
Molecular Function:
cell adhesion molecule
membrane-bound signaling molecule
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.