Invitrogen
Manufacturer Code:MA517937
Catalog # PIMA517937
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Armenian hamster |
Class | Monoclonal |
Type | Antibody |
Clone | Eat2 |
Immunogen | Mouse CD81 (TAPA-1) |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Armenian Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: 26 kDa cell surface protein TAPA-1; 26 kDa cell surface protein TAPA-1 CD81 antigen CD81 antigen target of antiproliferative antibody 1 tetraspanin-28 tspan-28; CD 81 antigen; CD81; CD81 antigen; CD81 antigen (target of antiproliferative antibody 1); Target of the antiproliferative antibody 1
Gene Aliases: Cd81; Tapa-1; Tapa1; Tspan28
UniProt ID: (Mouse) P35762
Entrez Gene ID: (Rat) 25621, (Mouse) 12520
Molecular Function: cell adhesion molecule membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.