R&D Systems
Manufacturer Code:MAB46151SP
Catalog # MAB46151SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 454729 |
Immunogen | HEK293 human embryonic kidney cell line transfected with human CD81 Accession # P60033 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
Protein Aliases: 26 kDa cell surface protein TAPA-1; CD81; CD81 antigen; CD81 antigen (target of antiproliferative antibody 1); CD81 CD81 antigen CD81 antigen (target of antiproliferative 1) CD81 molecule CVID6 TAPA1 TAPA-1 Target of the antiproliferative 1 Tetraspanin-28 Tspan-28 TSPAN28 tspan-2826 kDa cell surface protein TAPA-1 TSPAN28S5.7; Target of the antiproliferative antibody 1; Tetraspanin-28; Tspan-28
Gene Aliases: CD81; CVID6; S5.7; TAPA1; TSPAN28
UniProt ID: (Human) P60033
Entrez Gene ID: (Human) 975
Molecular Function: cell adhesion molecule membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.