Invitrogen
Manufacturer Code:12080082
Catalog # 50-104-81
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3H5 |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: B7-1 B7.1 Cd28l Ly-53 Ly53 MIC17 TS/A-1 LAB7 B7.1 CD28LG CD28LG1; Cd80 a rat homolog of the human CD28/CTLA - 4 ligand (B7-1); CD80 antigen; T-lymphocyte activation antigen CD80
Gene Aliases: B7-1
Entrez Gene ID: (Rat) 25408
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.